Adapter, Vacuum Jacketed, Ace Glass Incorporated
Supplier: Ace Glass
5140-10
89051-342EA 708.93 USD
89051-342
Adapter, Vacuum Jacketed, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This vacuum jacketed adapter is used as a distilling head for connecting the top of the column with the side condenser.
  • Top [ST] inner joint
  • Side and bottom [ST] inner joints
  • Accommodates thermometer
  • Fabricated from borosilicate glass
The top joint is a standard taper 10/30 inner for a 76mm immersion thermometer. Side and bottom joints are standard taper 24/40 inner. Side arm has drip tip.