ThermoSafe® Medical Specimen Transporter Totes, Sonoco ThermoSafe
Supplier: Sonoco Thermosafe
640 932 641
89047-300CS 122.6 USD
89047-300 89049-956 89049-958 15713-614
ThermoSafe® Medical Specimen Transporter Totes, Sonoco ThermoSafe
Coolers Transport Coolers
Medical transporter totes for on-site collection, storage, and transportation of temperature-sensitive specimens, organs, medications, and medical devices.
  • For on-site collection and specimen transport
  • Durable and washable insulated carriers
  • Validated for temporary storage
Totes are made of durable, washable and weather-resistant nylon, with a stain- and odor-resistant expanded polypropylene (EPP) insulated interior
Non-qualified small and large locking totes are for short-term storage and transport of temperature-sensitive tissues, serums,More Product Information