VWR® Mortars and Pestles, Agate
Supplier: VWR International
C-100 C-101 C-102 C-103 C-104 C-105 C-106 C-107 C-108 C-109 C-110 C-111 C-112 C-113 C-115 C-116 C-117
89037-480CS 2325.51 USD
89037-480 89037-482 89037-484 89037-486 89037-488 89037-490 89037-492 89037-494 89037-496 89037-498 89037-500 89037-502 89037-504 89037-506 89037-510 89037-512 89037-514
VWR® Mortars and Pestles, Agate
Mortars and Pestles Mortars with Pestles
Mortars and pestles are made of thick-walled agate, which is the most common variety of chalcedony, a semiprecious, very hard silica mineral.
  • Thick-walled agate construction
  • Highly polished surfaces
  • Octagonal shape
They feature highly polished, glossy grinding surfaces. Mortar is octagonal in shape.
Ordering information: Supplied in mortar and pestle sets or pestles only.
Order Now
SPECIFICATIONS
Environmentally Preferable