VWR® Critical Swab® Slim Cotton Double Tipped Swab, Fine Paper Handle
Supplier: VWR International
89031-292
89031-292CS 304.73 USD
89031-292
VWR® Critical Swab® Slim Cotton Double Tipped Swab, Fine Paper Handle
Swabs Swabs for Sampling
Swab features a fine paper stem and two slim tips composed of lintless, highly absorbent cotton.
  • Antistatic packing
  • Lintless, highly absorbent cotton
Made in the USA.
Packaged in an antistatic leaf pack.
Certifications: CE marked.
Ordering information: Packaged as 50 leaves of 25 swabs per leave.