Powder Funnels, PTFE, Chemglass
Supplier: Chemglass
CG-1764-01 CG-1764-02 CG-1764-03 CG-1764-04 CG-1764-05
89028-766EA 41.93 USD
89028-766 89028-768 89028-770 89028-772 89028-774
Powder Funnels, PTFE, Chemglass
Funnels Powder Funnels
PTFE powder funnels feature a non-stick internal finish.
  • Made of PTFE
  • Chemically inert with super