Automation Pipette Tips for PatchXpress Workstations, Thermo Scientific
Supplier: Thermo Scientific
902-261XP
89025-942CS 227.68 USD
89025-942
Automation Pipette Tips for PatchXpress Workstations, Thermo Scientific
Pipette Tips
Thermo Scientific provides a solution for your PatchXpress robotic pipette tips.
Our low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot. Each lot must pass a 15-point QC inspection before tips are released. All tips are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.