CoolSafe PCR Tube Cooling Chamber, Diversified Biotech
Supplier: Diversified Biotech
CHAM-1020
89024-892EA 1626.9 USD
89024-892
CoolSafe PCR Tube Cooling Chamber, Diversified Biotech
Coolers Benchtop Coolers
Anodized aluminum cooling chamber with 212 wells is ideal for temperature-sensitive liquid transfers
May be used when storing samples in ice buckets, freezers, and on hot plates. The chamber accommodates two 96-well PCR trays or 192 tubes sized 0.2mL, as well as twelve 0.5mL tubes and eight 1.5mL tubes.