Desiccant in Tyvek® Pouch, SCS
Supplier: Desco
1/6PLDES1200 1PLDES300
89024-376PK 388.67 USD
89024-376 89024-380 89024-382
Desiccant in Tyvek® Pouch, SCS
Desiccators Desiccants
This efficient desiccant begins working by absorbing airborne moisture left inside a bag when vacuum packaging is complete. It effectively captures moisture that manages to pass through the bag material.
  • Provides protection for moisture sensitive electronics (such as Surface Mount Technology)
  • Best practice is to use with a Humidity Indicator Card
  • Recommended amount of desiccant is dependent on the interior surface area of the bag to be used
  • Can be baked and re-used
  • No hazardous substances; see RoHS 3, REACH, Toxins in Packaging Statement
Secured in aMore Product Information