Dri-Shield® Moisture Barrier Bags, 3000 Series, SCS, Desco Industries
Supplier: Desco
D301020 D301030 D301618
89024-370PK 288.03 USD
89024-370 89024-372 89024-374
Dri-Shield® Moisture Barrier Bags, 3000 Series, SCS, Desco Industries
Bags Storage Bags
This foil moisture barrier bag provides protection for ESD sensitive and moisture sensitive electronics, typically SMT (Surface Mount Technology) items. Best practice is to use with Desiccant and a Humidity Indicator Card. A nylon layer helps to strengthen the bag. The bags are heat sealable and suitable for vacuum packaging. They are printed with ESD protective and moisture sensitive warning symbols, and a lot code for traceability.
  • 0.006" (6 mil) thick bag constructed from layers of nylon, foil, and polyethylene
  • Nylon reinforcement layer is an ideal
More Product Information