Dri-Shield® Moisture Barrier Bags, 2000 Series, SCS, Desco Industries
Supplier: Desco
700810 7001012 7001020 7001216 7001518 7001824
89024-326PK 60.23 USD
89024-326 89024-330 89024-332 89024-338 89024-344 89024-352
Dri-Shield® Moisture Barrier Bags, 2000 Series, SCS, Desco Industries
Bags Storage Bags
This aluminized moisture barrier bag is designed to provide packaging for ESD and moisture sensitive items, both inside and outside an ESD protected area. The bags are heat sealable and suitable for vacuum packaging.
  • 0.0036" (3.6 mil) thick bag film laminate constructed from multiple layers of aluminized polyester and polyethylene
  • Designed for moisture and electrostatic discharge (ESD) sensitive items (such as SMT)
  • < 0.03 Grams Moisture Vapor Transmission Rate (MVTR); designed for dry packing of moisture sensitive devices and to protect contents from
More Product Information