Static Shielding Bags 1000, Desco Industries
Supplier: Desco
10023 10024 10026 10048 100424 100426 10056 10069 100612 100614 100616 100618 100624 100626 1006.516 100710 100711 100715 100716 100818 100830 100912 1001026 1001114 1001214 1001225 1001430 1002020 1002224 1003024 30033 30035 30046 30048 300424 300430 300510 300610 300624 300630 300715 30088 3001010 3001024 3001030 3001418 3001624 3001818 3002024
89024-028PK 9.11 USD
89024-028 89024-030 89024-032 89024-038 89024-040 89024-042 89024-044 89024-048 89024-056 89024-058 89024-062 89024-064 89024-066 89024-068 89024-070 89024-072 89024-078 89024-082 89024-084 89024-086 89024-088 89024-092 89024-100 89024-104 89024-106 89024-110 89024-114 89024-118 89024-122 89024-126 89024-128 89024-136 89024-146 89024-164 89024-166 89024-170 89024-178 89024-182 89024-184 89024-188 89024-190 89024-192 89024-194 89024-198 89024-202 89024-204 89024-206 89024-208 89024-210 89024-218 89024-224 89024-226 89024-232 89024-236 89024-240 89024-242 89024-246 89024-248
Static Shielding Bags 1000, Desco Industries
Bags Storage Bags
These transparent metal-in static shielding bags are designed to provide a static safe environment for sensitive electronic devices.
  • Bags are printed with an ESD warning symbol and a lot code for traceability
  • Open top and ZipTop styles are available
  • Bags are heat sealable
These bags are manufactured from a polyester, metal, polyethylene laminate. The polyester dielectric in concert with the metal layer is intended to provide shielding of Electrostatic Discharge (ESD) and to help minimize the penetration of electric field.
Certifications: These bagsMore Product Information