SAF-T-FIT™ Plus N1125 Disposable Respirators, Honeywell Safety
Supplier: Honeywell Safety Products
14110394
89013-612BX 110.08 USD
89013-612 89013-604 89013-616 89013-610 89013-600 89013-614 89013-602
SAF-T-FIT™ Plus N1125 Disposable Respirators, Honeywell Safety
Respirators Disposable Respirators Particulate Respirators
Ideal for welding, soldering, brazing, torch cutting, metal pouring, grinding, sanding, sweeping, or other hot and dusty operations. Respirators feature an adjustable boomerang nose seal for a secure fit and added comfort. Nose seal is color-coded for easy size recognition. Respirators are equipped with an exhalation valve for ease of breathing and exceptional comfort during extended wear.
  • Offers latex-free N95 protection from particulate aerosols free of oil
  • Exhalation valve improves comfort during extended wear
  • Adjustable boomerang nose seal fits
More Product Information