Howard Leight SmartFit® Reusable Earplugs, Honeywell Safety
Supplier: Honeywell Safety Products
SMF-30 SDT-30
82007-557CS 2465.53 USD
82007-557 89008-849
Howard Leight SmartFit® Reusable Earplugs, Honeywell Safety
Earplugs Reusable Earplugs
Ideal for applications requiring constant hearing protection over extended periods of time, such as manufacturing-line operations, petroleum refining, and mining
  • Material Technology™ contours to ear canal for better fit
  • Return to normal shape when removed
  • Flat triple-fin design maximizes contact in ear canal
  • Flanged handle for easy insertion/removal and high visibility for compliance checks
  • Hygienic design, cleans easily
  • One size fits most users
Standard earplugs are orange and include a detachable nylon cord. Detectable earplugs are blue and featureMore Product Information