Hydrovoid Atmosphere Control System, Air Control
Supplier: Air Control
5330
89008-254EA 6841.31 USD
89008-254
Hydrovoid Atmosphere Control System, Air Control
Glove Boxes and Glove Bags
Economical and durable glove box control system is ideal for general laboratory manipulation requiring dry or special inert atmospheres
Fabricated of optically clear acrylic, unit allows the operator unhampered visibility within the main chamber and the air lock for optimum performance. The sloping front offers operator comfort. The solvent-fused seams provide an airtight enclosure for protection of the operator and process. Unit features closed-cell neoprene gasketing around doors, 120V electrical receptacles, and high-density polyethylene arm ports with adjustable rings.
Order Now
SPECIFICATIONS
Environmentally Preferable