E-A-Rsoft™ Metal Detectable Corded Earplugs, 3M™
Supplier: 3M
7000029952
89006-808BX 237.6 USD
89006-808
E-A-Rsoft™ Metal Detectable Corded Earplugs, 3M™
Earplugs Reusable Earplugs
3M™ E-A-Rsoft™ metal detectable earplugs were specially developed for food processing and other industries where it is critical to keep earplugs from contaminating products and processes.
Recognized by most metal-sensing systems, these earplugs feature an implanted stainless steel ball bearing in the plug and metal particles in the cord.
Order Now
SPECIFICATIONS
Environmentally Preferable