VWR® Sample Bags for the Seward Stomacher® Blender
Supplier: VWR International
11216-890 11216-892 11216-898 11216-900 11216-902 89003-820 89085-562 89085-564 89085-566 10048-880 10048-882 10048-884 10048-886 10048-888 10048-890 76200-884
11216-890CS 589.84 USD
11216-890 11216-892 11216-898 11216-900 11216-902 89003-820 89085-562 89085-564 89085-566 10048-880 10048-882 10048-884 10048-886 10048-888 10048-890 76200-884
VWR® Sample Bags for the Seward Stomacher® Blender
Bags Blender Bags
Sample bags are designed to withstand the paddle action of Stomacher® lab blenders.
  • Pure or hybrid polyethylene construction
  • Highly transparent
  • Suitable for general-purpose blending or analysis testing
  • Some bags feature write-on strip
  • Malleable yet durable
FILTRA-BAG® bags are made of a mix of polyethylene and nylon.
SECURE-T® bags are made of heat extruded polyethylene tubing for internal sterility and to eliminate the need for side seals. Bags are malleable yet durable, with superior wall strength. May be used in general-purpose blending orMore Product Information