ceramus™-classic Bottle-Top Dispensers, Hirschmann
Supplier: Hirschmann
931000010 931110010 932000010 932110010 933000010 933110010 934000010 934110010 9350000 9351100 9361100 9360000
83007-610EA 910.59 USD
83007-610 83007-612 83007-614 83007-616 83007-618 83007-620 83007-622 83007-624 83007-626 83007-628 83007-632 83007-630
ceramus™-classic Bottle-Top Dispensers, Hirschmann
Bottle Top Dispensers
Dispensers feature a ceramic plunger that is made of solid, 99.7% pure aluminum oxide, which guarantees the highest degree of chemical resistance, without abrasion or swelling
The cylinder is made of calibrated DURAN® (borosilicate 3.3) glass, is predominantly inert and wear-free, and is protected by robust plastic sheathing. The protective sheathing incorporates two glass windows so that air bubbles can be easily identified. The check valves are also predominantly inert and wear-free. The ball and seat are made of borosilicate glass, the valve springs are made of Hastelloy® material, theMore Product Information