96-Well Plate Lids, Greiner bio-one
Supplier: Greiner Bio-One
691161 656101 656161 656170 656171 656172 656176 656178 656190 656191 656199 691101 656197
82050-068CS 236.08 USD
82050-068 82050-823 82050-825 82050-827 82050-829 82050-830 82050-832 82050-834 82050-836 82050-838 82050-840 82051-582 89131-671
96-Well Plate Lids, Greiner bio-one
Lids Microplate Lids
Made of polycarbonate or polystyrene resin, these lids are best suited for chimney-well style 96-well plates in cell culture applications.
  • Polycarbonate or polystyrene resin construction
  • Fully skirted lids
  • Free of RNase and DNase
  • Nonpyrogenic
Half-area plates maintain ANSI/SBS standards and well-to-well pitch, yet feature a well diameter design that offers one-half the well surface area of standard plates
Standard full skirt lids allow easy gripping of the lid manually or robotically. Low-profile lids feature a partial skirt, facilitating roboticMore Product Information