TRUTOL® Glucose Tolerance Beverages, NERL Diagnostics
Supplier: Thermo Fisher Scientific
401009P 401503P 401025P 401272P 401074P 401576P 401223P 401526P 401207P
82028-504CS 375.39 USD
82028-504 82028-506 82028-512 82028-516 82028-520 82028-522 82028-508 82028-514 82028-500
TRUTOL® Glucose Tolerance Beverages, NERL Diagnostics
Beverages
Caffeine-free TRUTOL® beverages are used to screen for hyperglycemia (diabetes mellitus and gestational diabetes) and hypoglycemia (low blood sugar).
  • Screening entails administering a precise dose of glucose and then testing the level of glucose in the patient’s blood at specific time intervals
  • Bottles feature volume graduations imprinted on the label to aid in dispensing smaller quantities
Bottle size: 296 ml (10 oz.).
Certifications: Manufactured to meet FDA standards. Certified kosher.
Ordering information: Carbonated flavors are packaged inMore Product Information