Proteomic Grade Detergent Solutions
Supplier: G-Biosciences
DG001 DG003 DG002 DG005 DG006 DG007 DG008 DG010 DG004 DG013 DG521 DG009 DG012 DG011
82022-966EA 211.78 USD
82022-966 82022-970 82022-968 82022-974 82022-976 82022-978 82022-980 82022-984 82022-972 82022-988 95057-526 82022-982 82022-986 82023-060
Proteomic Grade Detergent Solutions
Cleaning Agents Detergents and Surface Cleansers
Many commercial grade detergents contain elevated levels of sulfhydryl oxidizing agents, peroxides, salts and carbonyl compounds.
Our Proteomic Grade Detergent Solutions contain reduced peroxides and carbonyl compounds. In addition, the detergents have less than 50 μS conductivity. These detergents are offered as 10% aqueous solutions, sealed under inert gas and are suitable for protein applications. These non-ionic detergents are suitable for isolating membrane-protein complexes.
G-Biosciences Proteomic Grade Detergent Solutions include: Triton® X-100, Triton® X-114, Brij® 35, Brij® 58
More Product Information