Sulfo-DST (Sulfodisuccinimidyl tartrate)

Supplier: G-Biosciences

Synonyms: Sulfo DST, Bis(Sulphosuccinimidyl)tartrate, Bis(Sulfosuccinimidyl)tartrate

BC06
82022-780EA 458.51 USD
82022-780
Sulfo-DST (Sulfodisuccinimidyl tartrate)
Sulfo-DST (Sulfodisuccinimidyl tartrate)

Sulfodisuccinimidyl tartrate Homobifunctional Cross Linker.

Formula: C₁₂H₁₂N₂O₁₆S₂
MW: 504.357 g/mol
CAS Number: 118674-04-7

Order Now

Specification Test Results

Spacer Arm (Å) 6.4
Reactive Toward Primary Amines
Membrane Permeable No
Water Soluble Yes
Cleavable/ Reversible Oxidizing Agents (Periodate)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR