FOCUS™ Cytoplasmic and Nuclear, G-Biosciences
Supplier: G-Biosciences
786-248
82022-674EA 667.78 USD
82022-674
FOCUS™ Cytoplasmic and Nuclear, G-Biosciences
Protein Extraction Protein Extraction Kits
Supplied with a strong chaotropic extraction buffer to solubilize both cytoplasmic and nuclear proteins, which is fully compatible with 2D gel electrophoresis.
G-Biosciences' FOCUS™ Cytoplasmic & Nuclear proteins fractionation kit is based on the Nuclear & Cytoplasmic Extraction kit (see figure), however it has been modified to be fully compatible with 2D electrophoresis and subsequent downstream processes.
The kit is provided with reagents necessary for fractionation of cytoplasmic and nuclear proteins as well as solubilization buffer suitable for IEF/2D gels for better spot resolution.
Designed for >50 preps, depending on the tissue used, where one prep is 20x10^6 mammalian cells or 100 mg mammalian tissue.
Order Now
SPECIFICATIONS
Environmentally Preferable