Coventry™ ESD Swab, Pointed Foam Head, ITW Chemtronics®
Supplier: ITW Chemtronics
44070ESD
82021-218CS 1560.27 USD
82021-218
Coventry™ ESD Swab, Pointed Foam Head, ITW Chemtronics®
Swabs Swabs for Cleaning
Designed to eliminate damage from static electricity, the handle of this swab will dissipate 99% of a 5000V charge in less than two seconds
The plastic alloy used to make the handle is inherently static dissipative and contains no additives, coatings, or surfactants. The swab functions at any level of humidity and is highly resistant to solvents, including acetone. Swab features an ultraclean polyurethane foam head.