Monoject™ SoftPack Syringes, Sterile, Covidien
Supplier: Covidien
1181621100 1180320112 1182000555 1183500777 1180300777 1180321100 1180323100 1180325114 1180600777 1180300555 1180320100 1180321112 1180325058 1180600555 1181622112 1183500555 1183500888 1180322100 1180322112 1180327114 1182000777 1181621112 1181620112
82002-299CS 423.57 USD
82002-299 82002-252 82002-296 82002-304 82002-310 82002-246 82002-254 82002-264 82002-272 14222-240 82002-244 82002-250 82002-256 82002-268 82002-278 82002-302 82002-308 82002-312 82002-258 82002-260 82002-274 82002-306 82002-301 82002-297
Monoject™ SoftPack Syringes, Sterile, Covidien
Syringes Syringes, Medical
These latex-free syringes feature a one-step peel-away package that facilitates quick and easy opening for aseptic handling.
The 3mL syringes feature extended finger flanges and a longer plunger rod for greater control. The 3mL, 6mL, and 12mL syringes feature bold graduations; the 35mL and 60mL syringes feature bold dual graduations. The 35mL syringes also feature 1oz. graduations in 0.25oz. increments, and the 60mL syringes have 2oz. graduations in 0.25oz. increments.
Each individual package contains a color-coded gauge stripe and bar code for easy gauge identification. SyringesMore Product Information