Sodium carbonate (Soda ash dense) dense

Warning

Synonyms: Soda, Carbonic acid sodium salt, Calcined soda, Disodium carbonate, Carbonic acid disodium salt, Na-X, Bisodium carbonate, Carbonic acid, disodium salt, Soda ash

SAD50-050B
80700-081EA 51.36 USD
80700-081
Sodium carbonate (Soda ash dense) dense
Sodium carbonate
Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600
Melting Pt: 854
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
Merck Index: 14,08596

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR