PROTEIN GRADE® Detergent, C12E9, Calbiochem®, MilliporeSigma
Supplier: MilliporeSigma
205534-1SET
80600-992EA 406.46 USD
80600-992
PROTEIN GRADE® Detergent, C12E9, Calbiochem®, MilliporeSigma
Protein Extraction Protein Extraction Reagents
A non-ionic detergent used for the solubilization of membrane-bound proteins in their native state.
  • HLB number: 13.6
  • CMC: 80 μM
Packaging: Packaged under inert gas.