Poulten and Graf Fortuna™ Glass Syringes, Air-Tite Products
Supplier: Air-Tite
GT100 GL100 GT1 GT2 GT3 GT5 GT10 GT20 GT30 GT50 GL1 GL2 GL3 GL5 GL10 GL20 GL30 GL50
80089-560EA 119.16 USD
80089-560 80089-578 80089-544 80089-546 80089-548 80089-550 80089-552 80089-554 80089-556 80089-558 80089-562 80089-564 80089-566 80089-568 80089-570 80089-572 80089-574 80089-576
Poulten and Graf Fortuna™ Glass Syringes, Air-Tite Products
Syringes Syringes, Medical
Fortuna Optima syringes are specialty syringes made from very fine borosilicate glass.
  • Borosilicate glass
  • Interchangeable pistons and barrels
  • Available with Luer Slip Tip or Luer Lock Tip
These syringes are manufactured in Germany from very fine borosilicate glass with interchangeable pistons and barrels. They feature amber graduations and have Luer lock and slip tips.
Certifications: Meets ISO Standards.
Ordering information: Individually boxed.
Order Now
SPECIFICATIONS
Environmentally Preferable