SP Wilmad-LabGlass Long-Tip Sample Transfer NMR Pipettes, SP Industries
Supplier: SP Industries
803A
80087-518PK 273.32 USD
80087-518
SP Wilmad-LabGlass Long-Tip Sample Transfer NMR Pipettes, SP Industries
Tubes NMR Tubes
Wilmad Long-Tip Sample Transfer NMR Pipettes are designed for easy transfer of liquid samples contained in 5 mm Ext.Ø NMR tubes, long neck volumetric flasks or chromatography columns.
  • Manufactured from high quality ASTM Type 1 Class A glass
  • Resistant to most organic solvents
  • Transparency provides easy control of sample loading
  • Easily attach with a latex bulb (804), sold separately
  • Manufactured in a clean room
Free from organic and inorganic contamination. Special shaping process ensures a smooth surface to minimize sample loss.