Phenolic Screw Caps, Black, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
W242711
80090-042CS 199.23 USD
80090-042 80090-044 80076-568
Phenolic Screw Caps, Black, Wheaton
Closures Bottle Closures
Black phenolic screw caps feature a white rubber liner.
  • Chemical resistant
  • Temperature tolerant
These black phenolic screw caps are made of phenolic resin.
Caps provide resistance to most chemicals and are temperature tolerant.