Distilling Columns, Hemple, Chemglass
Supplier: Chemglass
CG-1228-11 CG-1228-05 CG-1228-08 CG-1228-09
80067-486EA 146.23 USD
80067-486 80067-478 80067-480 80067-482
Distilling Columns, Hemple, Chemglass
Distillation Components Distillation Columns
Distillations feature a row of indentations.
  • Plain distillation column
  • Includes a row of indentations to support column packing material
Column length listed is for the straight portion above the row of indentations and below the joint. Column has a standard taper top outer and lower inner joint.
Order Now
SPECIFICATIONS
Environmentally Preferable