Airfree® Schlenk Drying Chambers, Chemglass
Supplier: Chemglass
AF-0556-01 AF-0556-02 AF-0556-03 AF-0556-04 AF-0556-05 AF-0556-06
80064-956EA 592.67 USD
80064-956 80064-958 80064-960 80064-962 80064-964 80064-966
Airfree® Schlenk Drying Chambers, Chemglass
Baths Oil Baths
Chambers are designed for use in drying small quantities of material in dram vials.
  • Top adapter has a 001504mm Chem-Vac™ Chem-Cap® valve
  • Supplied with corresponding stainless steel pinch clamp and one Viton O-Ring
  • #40 O-Ring lower vessel overall dimensions: 40×100 mm (I.D.×H)
  • #50 O-Ring lower vessel overall dimensions: 50×100 mm (I.D.×H)
Chambers can also be used in the transfer of materials from glovebox to glovebox, or to the vacuum line. Lower vessel has a flat bottom permitting it to be stood upright on the bench or in a glovebox. Complete item isMore Product Information