Vacuum Trap, Solvent Vapor, Plastic-Coated, Chemglass
Supplier: Chemglass
CG-4519-01
80063-072EA 1827.79 USD
80063-072
Vacuum Trap, Solvent Vapor, Plastic-Coated, Chemglass
Vacuum Traps
Trap is designed for large volume solvent vapor removal during rotary evaporation.
Trap is placed between the vacuum source and the rotary evaporator to prevent solvent vapors from entering the vacuum pump. Plastic-coated to contain solvent in case of breakage and supplied complete with a UHMW lid with handle.
Order Now
SPECIFICATIONS
Environmentally Preferable