4-Methylumbelliferyl-alpha-D-glucopyranoside

Supplier: MilliporeSigma

Synonyms: 4-Methyl-7-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one, 4-Methylumbelliferyl-α-D-Glucopyranoside, 4-MU-alpha-D-Glc

474426-100MG
80058-382EA 207.34 USD
80058-382
4-Methylumbelliferyl-alpha-D-glucopyranoside
4-Methylumbelliferyl-alpha-D-glucopyranoside
Formula: C₁₆H₁₈O₈
MDL Number: MFCD00067661
CAS Number: 17833-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR