D-(+)-Mannose

Supplier: MilliporeSigma

Synonyms: (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal

4440-100GM
80058-332EA 217.24 USD
80058-332
D-(+)-Mannose
D(+)-Mannose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 376
Melting Pt: 133
MDL Number: MFCD00064122
CAS Number: 3458-28-4
Merck Index: 13,05772

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR