Glycine

Supplier: MilliporeSigma

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

3570-1KG 3570-500GM
80058-140EA 95.28 USD
80058-140 80058-142
Glycine
Glycine
Formula: NH₂CH₂COOH
MW: 75.07 g/mol
Boiling Pt: 181
Melting Pt: 83
MDL Number: MFCD00008131
CAS Number: 56-40-6
Merck Index: 13,04504

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR