Chlorpromazine hydrochloride, Calbiochem®

Supplier: MilliporeSigma

Synonyms: 2-Chloro-10-[3-(dimethylamino)-1-propyl]phenothiazine Hydrochloride

215921-500MG
80055-146EA 129.14 USD
80055-146
Chlorpromazine hydrochloride, Calbiochem®
Chlorpromazine hydrochloride

Inhibits calmodulin-dependent stimulation of cyclic nucleotide phosphodiesterase (IC50=17µM). Acts as a peripheral vasodilator. An inhibitor of lysosomal sphingomyelinase. An inhibitor of TNF-[alpha] production. Inhibits nitric oxide synthase (NOS) in mouse brain and prevents lipopolysaccharide induction of NOS in murine lung. Potent PLA2 inhibitor.

Formula: C₁₇H₁₉ClN₂S
MW: 355.32 g/mol
MDL Number: MFCD00012654
CAS Number: 69-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR