Human Thrombin (from Plasma)
Supplier: MilliporeSigma
605195-1000U 605195-100U 605195-5000U
80054-692EA 715.82 USD
80054-692 80054-694 80054-696
Human Thrombin (from Plasma)
Enzymes
Designed for use in thrombin clotting time tests.
Key enzyme that activates factor XIII and converts fibrinogen into fibrin. Soluble in aqueous buffers or water. Lyophilized from 200 mm NaCl, 50 mm sodium citrate, 0.1% PEG, pH 6.5.