β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) ≥95% (by enzymatic purity)

Supplier: MilliporeSigma

Synonyms: Nicotinamide adenine dinucleotide reduced, disodium salt (reduced form), NADH disodium salt, ((2R,3S,4R,5R)-5-(6-Aminopurin-9-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy-((((2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy)hydroxyphosphoryl)oxyphosphinic acid disodium salt

481913-500MG 481913-1GM
80053-328EA 125.15 USD
80053-328 80053-326
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) ≥95% (by enzymatic purity)
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form)

Functions as a coenzyme of a large number of oxidoreductases.


  • Yellow solid
  • Soluble in H₂O

Formula: C₂₁H₂₇N₇Na₂O₁₄P₂
MW: 709.41 g/mol
MDL Number: MFCD00036200
CAS Number: 606-68-8

Order Now

Specification Test Results

Synonyms β-Nicotinamide-adenine Dinucleotide, Reduced, 2Na
Form Yellow Solid
Purity ≥95% by enzymatic assay (dry basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR