Fura-2 LR/AM ≥95% (by HPLC), Calbiochem®

Supplier: MilliporeSigma

Synonyms: Fura-2 Leakage Resistant (AM)

344911-1SET
80051-796EA 415.12 USD
80051-796
Fura-2 LR/AM ≥95% (by HPLC), Calbiochem®
Fura-2 LR/AM

Acetoxymethyl (AM) derivative of FURA-PE3 (80051-794) with spectral properties similar to the parent compound


Can be loaded into cells in a manner similar to FURA 2/AM (80051-788). Hydrolysis by non-specific esterases traps the indicator in the cytosol. Excitation maximum: 364nm (low Ca2+) and 335nm (high Ca2+); emission maximum: 508nm (low Ca2+) and 495nm (high Ca2+).

Formula: C₅₅H₆₃N₅O₂₉
CAS Number: 172890-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR