Caspase-1 Substrate X, Fluorogenic, Calbiochem®

Supplier: MilliporeSigma

218739-5MG
80050-782EA 269.29 USD
80050-782
Caspase-1 Substrate X, Fluorogenic, Calbiochem®
Electrophoresis Reagents Western Blotting Reagents

A fluorogenic substrate for detecting caspase-1 (ICE), caspase-4, and caspase-5 activity


Excitation maximum: ~365–380nm; emission maximum: ~430–460nm.

Order Now

SPECIFICATIONS

Environmentally Preferable
Form Lyophilized
Molecular Weight 784.8
Purity 95% by HPLC
Solubility DMSO

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR