Transfection Reagent, 293-FREE, Novagen, MilliporeSigma
Supplier: MilliporeSigma
72181-4 72181-5 72181-3
80018-036EA 2289.56 USD
80018-036 80018-038 80018-034
Transfection Reagent, 293-FREE, Novagen, MilliporeSigma
Transfection Reagents
Has a unique polycationic liposomal formulation and is designed expressly for transfection of HEK 293 cells grown in suspension culture
Derived from non-animal sources. Provided as a sterile, ready-to use solution.
Order Now
SPECIFICATIONS
Cell Specific Mammalian, Insect
Environmentally Preferable
Bioproduction Yes
DNA Yes