2xYT Broth Dehydrated Culture Media, MilliporeSigma
Supplier: MilliporeSigma
71755-4
80017-506EA 900.81 USD
80017-506
2xYT Broth Dehydrated Culture Media, MilliporeSigma
Media for Microbiology Dehydrated Media for Microbiology
Media is granulated.
  • Granulation preserves homogeneity
  • Easy to weigh out
  • Dissolves faster than powdered media
Granulation preserves homogeneity by capturing the media’s original component distribution in each granule and preventing settling out
2xYT Broth Contents: Tryptone 16 g/L; Yeast extract 10 g/L; NaCl 5 g/L (pH 6.8-7.2, at 25 °C).
Dangers associated with inhalation of fine dust and toxic substances are significantly reduced with granulated media. It is easier to weigh out and dissolves faster than powdered media.