CytoFLEX Mosaic Detection Modules
Supplier: Beckman Coulter
D19581 D19582 D19583 D19584 D19585 D19586 D19589 D20160 D20161 D20162 D20163 D20164 D20165 D20166
77886-964EA 260000 USD
77886-964 77886-966 77886-968 77886-970 77886-972 77886-974 77886-976 77886-978 77886-980 77886-982 77886-984 77886-986 77886-988 77886-990
CytoFLEX Mosaic Detection Modules
Cell Counters and Flow Cytometers Flow Cytometers
The CytoFLEX Mosaic Detection Module is the natural evolution of the CytoFLEX platform. A smart approach to spectral analysis, the detection module connects to the CytoFLEX LX/S flow cytometer, providing the flexibility to mix and match different detection methods in one instrument.
  • A flexible approach to spectral flow cytometry
With up to 88 detection channels, the CytoFLEX Mosaic Detection Module enhances the CytoFLEX S or LX flow cytometer, empowering users to tackle the most complex experiments. Together, they create the most advanced and powerful
More Product Information