2-(Aminomethyl)-6-(trifluoromethyl)pyridine ≥98%

Supplier: Ambeed
Danger

Synonyms: (6-(Trifluoromethyl)pyridin-2-yl)methanamine

A781770-1G A781770-250MG A781770-100MG
77627-552EA 515 USD
77627-552 77627-554 77878-631
2-(Aminomethyl)-6-(trifluoromethyl)pyridine ≥98%
2-(Aminomethyl)-6-(trifluoromethyl)pyridine

Caution: for Research Use Only

Formula: C₇H₇F₃N₂
MW: 176.139 g/mol
MDL Number: MFCD10696232
CAS Number: 916304-19-3
UN: 2735
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR