AZD0780 98%

Supplier: Ambeed
Warning

Synonyms: 6'-(((1S,3S)-3-((5-(Difluoromethoxy)pyrimidin-2-yl)amino)cyclopentyl)amino)-2H-[1,3'-bipyridin]-2-one

A1937165-1MG A1937165-100MG A1937165-10MG A1937165-25MG A1937165-5MG
77625-942EA 97.85 USD
77625-942 77877-482 77877-483 77877-484 77877-485
AZD0780 98%
AZD0780

Caution: for Research Use Only

Formula: C₂₀H₂₀F₂N₆O₂
MW: 414.41 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34676592
CAS Number: 2455427-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR