Frontier™ 5000 Multi-Pro Centrifuges, 120 V
Supplier: Ohaus
30314811 30314813 30314815 30314817 30314819 30553037 30553103 83041046 83041048
77874-024EA 4184.13 USD
77874-024 77874-026 77874-028 77874-030 77874-032 77874-106 77874-108 77918-524 77918-526
Frontier™ 5000 Multi-Pro Centrifuges, 120 V
Centrifuges Benchtop Centrifuges
Frontier™ 5000 Multi-Pro Centrifuges are designed for universal use in multiple applications in research, industrial, and clinical laboratories and can be combined with our range of rotors and accessories for customized use. Available in 11 models with capacities from 0.2 ml to 4×1000 ml, they feature high-speed performance at maximum volume, automatic rotor identification, and safety features.
  • Corrosion-resistant stainless steel chamber with Polycarbonate/ABS plastic lid
  • Offer a high-speed centrifugation platform which can be customized to fit workflow
More Product Information