3,5-Di-tert-butyl-2-hydroxybenzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Di-tert-butylsalicylaldehyde

A140025-100G A140025-25G A140025-500G
77027-448EA 17 USD
77027-448 77027-450 77027-452 77871-673
3,5-Di-tert-butyl-2-hydroxybenzaldehyde 97%
3,5-Di-tert-butyl-2-hydroxybenzaldehyde

Caution: for Research Use Only

Formula: C₁₅H₂₂O₂
MW: 234.34 g/mol
MDL Number: MFCD00191998
CAS Number: 37942-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR