Orion™ Pro Star DO213 Dissolved Oxygen Bench Meters
Supplier: Thermo Fisher Scientific
PSTAR2130 PSTAR2135 PSTAR2136 PSTAR2137
77867-998EA 1726 USD
77867-998 77868-000 77868-002 77868-004
Orion™ Pro Star DO213 Dissolved Oxygen Bench Meters
Electrochemistry Meters Oxygen Meters
The Orion™ Pro Star DO213 dissolved oxygen bench meter is designed to provide accurate, reliable electrochemistry testing. Featuring an intuitive interface, enhanced data reporting and robust functionality, the Orion™ Pro Star DO213 meter delivers advanced performance in a modern and simplified package.
  • Responsive capacitive touchscreen operable with gloves
  • Angled display to help ensure ergonomic viewing
  • Streamlined, easy to operate user interface
  • Vertical meter orientation helps optimize bench space
  • Supports GLP compliance
  • Stores 20 calibration logs per
More Product Information