Methyl 5-nitro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 95%

Supplier: Ambeed
Warning

A734975-250MG A734975-5G A734975-100MG
76652-038EA 21 USD
76652-038 77783-815 77783-814
Methyl 5-nitro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 95%
Methyl 5-nitro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Caution: for Research Use Only

Formula: C₁₄H₁₈BNO₆
MW: 307.1068 g/mol
MDL Number: MFCD23379498
CAS Number: 1393477-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR