Bottle with Sprayer, Irradiated
Supplier: Contec
SB32IR
77776-828CS 155.4 USD
77776-828
Bottle with Sprayer, Irradiated
Bottles Spray Bottles
A convenient trigger sprayer with settings for stream or mist spraying is included with the 32 oz. bottles.
  • Each bottle has a lot number code and expiration date for easy record keeping
  • Product expires 5 years after date of manufacture
  • Validated sterile to 10⁻⁶ Sterility Assurance Level (SAL)
The stream setting is ideal for targeting specific hard to reach areas, while the mist setting is used to cover larger areas. Each leakproof bottle is induction sealed, capped, and shipped with an uninstalled trigger sprayer to eliminate the occurrence of leakage.More Product Information